Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fbx32 Rabbit anti-Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206162
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6162 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6162 Supplier Fabgennix Inc Supplier No. ATR101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fbx32 Polyclonal antibody specifically detects Fbx32 in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Fbx32
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene FBXO32
Gene Accession No. Q91Z62, Q9CPU7
Gene Alias 4833442G10Rik; AI430017; atrogin 1; atrogin1; atrogin-1; atrophy gene 1; F-box only protein 32; F-box protein 32; Fbx32; fbxo25; FBXO32; FLJ32424; Gm20361; MAFbx; MGC108443; MGC137646; MGC33610; Muscle atrophy F-box protein; similar to F-box only protein 32 isoform 1; zgc:56479
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 240-280 on Atr1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171043, 67731
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.