Learn More
Invitrogen™ FBXL10 Polyclonal Antibody

Description
Peptide sequence: LSFFKRCGNI CHIDLRYCKQ VTKEGCEQFI AEMSVSVQFG QVEEKLLQKL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
Specifications
Specifications
| Antigen | FBXL10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | KDM2B |
| Gene Accession No. | Q8NHM5 |
| Gene Alias | [Histone-H3]-lysine-36 demethylase 1B; Cxxc2; CXXC-type zinc finger protein 2; Fbl10; F-box and leucine-rich repeat protein 10; F-box protein containing leucine repeats 10; F-box protein FBL10; F-box/LRR-repeat protein 10; Fbxl10; JEMMA (Jumonji domain, EMSY-interactor, methyltransferase motif) protein; Jhdm1b; JmjC domain-containing histone demethylation protein 1B; jumonji C domain-containing histone demethylase 1B; Jumonji domain-containing EMSY-interactor methyltransferase motif protein; Kdm2b; Kiaa3014; lysine (K)-specific demethylase 2B; lysine demethylase 2B; lysine-specific demethylase 2B; NDY1; PCCX2; Protein JEMMA; Protein-containing CXXC domain 2 |
| Gene Symbols | KDM2B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.