Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXL12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544980 Supplier Invitrogen™ Supplier No. PA544980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PTEILSSCLT MPKLRVLELQ GLGWEGQEAE KILCKGLPHC MVIVRACPKE Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 86%; Rat: 93%.

Members of the F-box protein family, such as FBXL12, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FBXL12
Gene Accession No. Q9NXK8
Gene Alias 3110048D16Rik; Fbl12; F-box and leucine rich repeat protein 12; F-box and leucine-rich repeat protein 12; F-box protein containing leucine repeats 12; F-box protein FBL12; F-box/LRR-repeat protein 12; FBXL12; FLJ20188; LOC617304 protein
Gene Symbols FBXL12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FBXL12 (aa 271-320).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54850
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.