Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXL12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589921 Supplier Invitrogen™ Supplier No. PA589921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATLVELPDS VLLEIFSYLP VRDRIRISRV CHRWKRLVDD RWLWRHVDLT LYTMRPKVMW HLLRRYMASR LHSLRMGGYL FSGSQAPQLS PALLRALGQK CPNLKRLCLH VADLSMVPIT SLPSTLRTLE LHSCEISMAW LHKQQDPTVL PLLECIVLDR VPAFRDEHLQ GLTRFRALRS LVLGGTYRVT ETGLDAGLQE LSYLQRLEVL GCTLSADSTL LAISRHLRDV RKIRLTVRGL SAPGLAVLEG MPALESLCLQ GPLVTPEMPS PTEILSSCLT MPKLRVLELQ GLGWEGQEAE KILCKGLPHC MVIVRACPKE SMDWWM; Positive Samples: NIH/3T3, SW480, 293T, A-431, HeLa.

Members of the F-box protein family, such as FBXL12, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FBXL12
Gene Accession No. Q9EPX5, Q9NXK8
Gene Alias 3110048D16Rik; Fbl12; F-box and leucine rich repeat protein 12; F-box and leucine-rich repeat protein 12; F-box protein containing leucine repeats 12; F-box protein FBL12; F-box/LRR-repeat protein 12; FBXL12; FLJ20188; LOC617304 protein
Gene Symbols FBXL12
Host Species Rabbit
Immunogen Recombinant protein (or fragments).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30843, 54850
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.