Learn More
Invitrogen™ FBXL16 Polyclonal Antibody

Description
A suggested positive control is human spleen tissue lysate. PA5-21094 can be used with blocking peptide PEP-1208.
Specifications
Specifications
| Antigen | FBXL16 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | FBXL16 |
| Gene Accession No. | A2RT62, Q5MJ12, Q8N461 |
| Gene Alias | BC042620; C16orf22; c380A1.1; c380A1.1 (novel protein); FBL16; F-box and leucine rich repeat protein 16; F-box and leucine-rich repeat protein 16; F-box/LRR-repeat protein 16; Fbxl16; Scirr1; spinal cord injury and regeneration related 1; spinal cord injury and regeneration-related protein 1 |
| Gene Symbols | FBXL16 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.