Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXL16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563291 Supplier Invitrogen™ Supplier No. PA563291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPRITDMALE YVACDLHRLE ELVLDRCVRI TDTGLSYLST MSSLRSLYLR WCCQVQDFGL KH Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

FBXL16 (F-box and leucine-rich repeat protein 16), a member of the F-box protein family, is a probable substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. It is characterized by an approximately 40-amino acid F-box motif. F-box proteins interact with SKP1 through the F-box, and they interact with ubiquitination targets through other protein interaction domains. FBXL16 is a novel E2F1-regulated gene in crucial cell cycle regulation pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXL16
Gene Accession No. Q8N461
Gene Alias BC042620; C16orf22; c380A1.1; c380A1.1 (novel protein); FBL16; F-box and leucine rich repeat protein 16; F-box and leucine-rich repeat protein 16; F-box/LRR-repeat protein 16; Fbxl16; Scirr1; spinal cord injury and regeneration related 1; spinal cord injury and regeneration-related protein 1
Gene Symbols FBXL16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBXL16. Recombinant protein control fragment (Product #RP-103253).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146330
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.