Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXL18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109255 Supplier Invitrogen™ Supplier No. PA5109255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: PC-3M, rat liver tissue, human liver tissue, mouse liver tissue, A549.

Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL18
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene Fbxl18
Gene Accession No. Q96ME1
Gene Alias AI427077; B130019G13Rik; C330021B20Rik; Fbl18; F-box and leucine rich repeat protein 18; F-box and leucine-rich repeat protein 18; F-box/LRR-repeat protein 18; FBXL18
Gene Symbols Fbxl18
Host Species Rabbit
Immunogen Synthetic peptide within Human FBXL18 aa 756-805.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80028
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.