Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXL19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565146 Supplier Invitrogen™ Supplier No. PA565146
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGVSKKQLMWL LNRLQGLQEL VLSGCSWLSV SALGSAPLPA LRLLDLRWIE DVKDSQLREL LLPPPDTKPG QTESRGRLQG VAELRLAGLE LTDASLRLLL RHAPQLSALD LSHCAHVGDP SVHLLT Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL19
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXL19
Gene Accession No. Q6PCT2
Gene Alias 6030430O11; BC059812; CXXC11; Fbl19; F-box and leucine rich repeat protein 19; F-box and leucine-rich repeat protein 19; F-box/LRR-repeat protein 19; FBXL19; JHDM1C; jumonji C domain-containing histone demethylase 1C
Gene Symbols FBXL19
Host Species Rabbit
Immunogen Recombinant Human FBXL19. Recombinant protein control fragment (Product #RP-105900).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54620
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.