Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXL20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562180
Description
Immunogen sequence: IGAHCPELVT LNLQTCLQIT DEGLIT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Members of the F-box protein family, such as Scrapper, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. Scrapper is selectively expressed in the brain, broadly expressed within the mouse CNS and is abundant at presynaptic membrane. Scrapper has orthologs in C. elegans, D. melanogaster, and mammals which suggests that it might function as an important membrane-localized E3 ligase in various species. Scrapper is a major presynaptic E3 ubiquitin ligase that acts through RIM1a via degradation and the ubiquitin-proteasome-system (UPS) pathway to critically regulate synaptic transmission. This identifies protein degradation as a mechanism for holding synaptic communication in check.
Specifications
FBXL20 | |
Polyclonal | |
Unconjugated | |
FBXL20 | |
2610511F20Rik; 4632423N09Rik; AI849362; AL117906; C86145; Fbl2; Fbl20; F-box and leucine rich repeat protein 20; F-box and leucine-rich repeat protein 20; F-box protein FBL2; F-box/LRR-repeat protein 20; F-box/LRR-repeat protein 2-like; Fbxl2; Fbxl20; mKIAA4147; Scr; scrapper | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84961 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96IG2 | |
FBXL20 | |
Recombinant protein corresponding to Human FBXL20. Recombinant protein control fragment (Product #RP-100021). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction