Learn More
Invitrogen™ FBXL20 Polyclonal Antibody

Description
A suggested positive control is A-20 cell lysate. PA5-20485 can be used with blocking peptide PEP-0605.
Specifications
Specifications
| Antigen | FBXL20 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | FBXL20 |
| Gene Accession No. | Q96IG2, Q9CZV8, Q9QZH7 |
| Gene Alias | 2610511F20Rik; 4632423N09Rik; AI849362; AL117906; C86145; Fbl2; Fbl20; F-box and leucine rich repeat protein 20; F-box and leucine-rich repeat protein 20; F-box protein FBL2; F-box/LRR-repeat protein 20; F-box/LRR-repeat protein 2-like; Fbxl2; Fbxl20; mKIAA4147; Scr; scrapper |
| Gene Symbols | FBXL20 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.