Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXL21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113518
Description
FBXL21 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
FBL3B is a 434 amino acid protein encoded by the human gene FBXL21. FBL3B contains one 40 amino acid F-box region, making it a member of the F-box family. FBL3B also contains three LRR (leucine-rich) repeats. F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein) type E3 ubiquitin ligase complex and are involved in substrate recognition and recruitment for ubiquitination. F-box proteins are members of a large family that regulates cell cycle, immune response, signaling cascades and developmental programs by targeting proteins, such as cyclins, cyclin-dependent kinase inhibitors, IkB-a and b-catenin, for degradation by the proteasome after ubiquitination. FBL3B is a substrate-recognition component of the SCF complex that interacts with Skp1 p19 and CUL-1. FBL3B is also associated with expression and regulation of circadian and cryptochrome proteins.
Specifications
| FBXL21 | |
| Polyclonal | |
| Unconjugated | |
| FBXL21 | |
| D630045D17Rik; Fbl21; FBL3B; F-box and leucine rich repeat protein 21 (gene/pseudogene); F-box and leucine-rich repeat protein 21; F-box and leucine-rich repeat protein 3 pseudogene; F-box and leucine-rich repeat protein 3, pseudogene 1; F-box and leucine-rich repeat protein 3B; F-box protein Fbl3b; F-box/LRR-repeat protein 21; F-box/LRR-repeat protein 3B; Fbxl21; Fbxl3b; Fbxl3p; Fbxl3-ps1; Psttm | |
| Rabbit | |
| Protein G | |
| RUO | |
| 213311, 26223 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8BFZ4, Q9UKT6 | |
| FBXL21 | |
| Recombinant Human F-box/LRR-repeat protein 21 protein (78-137AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction