Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXL22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562992 Supplier Invitrogen™ Supplier No. PA562992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERPSETPPAP GVSWGPPPPG APVVISVKQE EGKQGRTGRR SHRAAPPCGF ARTRVCPPTF PGADAFPQ Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 36%.

Members of the F-box protein family, such as FBXL22, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL22
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXL22
Gene Accession No. Q6P050
Gene Alias 1110004B15Rik; Fbl22; F-box and leucine rich repeat protein 22; F-box and leucine-rich protein 22; F-box and leucine-rich repeat protein 22; F-box/LRR-repeat protein 22; Fbxl22
Gene Symbols FBXL22
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBXL22. Recombinant protein control fragment (Product #RP-98192).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283807
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.