Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXL3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10053784
Description
FBXL3 Polyclonal specifically detects FBXL3 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
FBXL3 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FBL3AF-box and leucine-rich repeat protein 3AF-box protein Fbl3a, FBL3F-box/LRR-repeat protein 3, F-box and leucine-rich repeat protein 3, F-box/LRR-repeat protein 3A, FBXL3A | |
Goat | |
Affinity Purified | |
RUO | |
26224 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_036290.1 | |
FBXL3 | |
Peptide with sequence KRGGRDSDRNSSEE-C corresponding to N-Terminus according to NP_036290.1. | |
0.1 mg | |
Primary | |
Expected from sequence similarity: Mouse, Rat, Dog, Pig . | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction