Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522133
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522133 Supplier Invitrogen™ Supplier No. PA522133
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Molt-4, C2C12. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (95%), Sheep (100%), Rhesus Monkey (100%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs, which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains several tandem leucine-rich repeats and is localized in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene FBXL3
Gene Accession No. Q8C4V4, Q9UKT7
Gene Alias Afh; AU041772; AW212966; FBK; FBL3; Fbl3a; F-box and leucine rich repeat protein 3; F-box and leucine-rich repeat protein 3; F-box and leucine-rich repeat protein 3A; F-box protein Fbl3a; F-box/LRR-repeat protein 3; F-box/LRR-repeat protein 3A; FBXL3; Fbxl3a; Ovtm; Play68; Protein after-hours; protein overtime
Gene Symbols FBXL3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FBXL3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26224, 50789
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.