Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXL6 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001291
Description
FBXL6 Polyclonal specifically detects FBXL6 in Human, Mouse samples. It is validated for Peptide ELISA.Specifications
FBXL6 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FBL6A, F-box and leucine-rich repeat protein 6FBL6F-box protein FBL6, F-box/LRR-repeat protein 6, PP14630 | |
Goat | |
Affinity Purified | |
RUO | |
26233 | |
Human, Mouse | |
IgG |
ELISA | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:8000 | |
NP_036294 | |
FBXL6 | |
Peptide with sequence CLEQLLTSPSPS corresponding to C-Terminus according to NP_036294.1, NP_078831.3. | |
0.1 mg | |
Primary | |
This is expected to recognize reported human isoforms (NP_036294.1 and NP_078831.3). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction