Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXL7 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298943AF594
Description
FBXL7 Polyclonal specifically detects FBXL7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FBXL7 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FBL7FBL6F-box protein Fbl7, F-box and leucine-rich repeat protein 7F-box protein FBL6/FBL7, F-box/LRR-repeat protein 7, KIAA0840 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human FBXL7. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 23194 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction