Learn More
Invitrogen™ FBXL8 Monoclonal Antibody (2E1)

Description
Peptide Sequence: MAEPGEGLPE EVLALIFRHL SLRDRAAAAR VCRAWAAAAT CSAVWHDTKI SCECELEGML PPYLSACLDH IHNLRLEFEP SRKPSRRAAI ELLMVLAGRA PGLRGLRLEC RGEKPLFDAG RDVLEAVHAV CGAASQLRHL DLRRLSFTLD DALVLQAARS CPELHSLFLD NSTLVGSVGP GSVLELLEAC PRLRALGLHL ASLSHAILEA LAAPDRAPFA LLALRCACPE DARASPLPNE AWVALRRRHP GLAVELELEP ALPAESVTRV LQPAVPVAAL RLNLSGDTVG PVRFAAHHYA ATLCALEVRA AASAELNAAL EELAARCAAL REVHCFCVVS HSVLDAFRAH CPRLRTYTLK LTREPHPWRP TLVA.
Specifications
Specifications
| Antigen | FBXL8 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2E1 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FBXL8 |
| Gene Accession No. | Q96CD0 |
| Gene Alias | FBL8; F-box and leucine rich repeat protein 8; F-box and leucine-rich repeat protein 8; F-box protein FBL8; F-box/LRR-repeat protein 8; FBXL8; RGD1566420 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.