Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXL8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114203
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114203 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114203 Supplier Invitrogen™ Supplier No. PA5114203
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ECELEGMLPP YLSACLDHIH NLRLEFEPSR KPSRRAAIEL LMVLAGRAPG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 86%; Horse: 93%; Human: 100%; Mouse: 85%; Pig: 93%; Rabbit: 92%; Rat: 86%.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class. It shares 78% sequence identity with the mouse protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FBXL8
Gene Accession No. Q96CD0
Gene Alias FBL8; F-box and leucine rich repeat protein 8; F-box and leucine-rich repeat protein 8; F-box protein FBL8; F-box/LRR-repeat protein 8; FBXL8; RGD1566420
Gene Symbols FBXL8
Host Species Rabbit
Immunogen Immunogen sequence: ECELEGMLPP YLSACLDHIH NLRLEFEPSR KPSRRAAIEL LMVLAGRAPG (aa 53-102).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55336
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.