Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113506
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113506 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113506 Supplier Invitrogen™ Supplier No. PA5113506
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FBXO10 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Members of the F-box protein family, such as FBXO10, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO10
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FBXO10
Gene Accession No. Q9UK96
Gene Alias F-box only protein 10; F-box only protein 10; LOW QUALITY PROTEIN: F-box only protein 10; F-box protein 10; F-box protein Fbx10; FBX10; FBXO10; PRMT11
Gene Symbols FBXO10
Host Species Rabbit
Immunogen Recombinant Human F-box only protein 10 protein (248-400AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26267
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.