Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXO10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570046
Description
This target displays homology in the following species: Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Rat: 86%.
Members of the F-box protein family, such as FBXO10, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
Specifications
FBXO10 | |
Polyclonal | |
Unconjugated | |
FBXO10 | |
F-box only protein 10; F-box only protein 10; LOW QUALITY PROTEIN: F-box only protein 10; F-box protein 10; F-box protein Fbx10; FBX10; FBXO10; PRMT11 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26267 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UK96 | |
FBXO10 | |
synthetic peptide directed towards the middle region of human FBXO10. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction