Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO11 Antibody, Novus Biologicals™
SDP

Catalog No. NBP155066 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP155066 Supplier Novus Biologicals Supplier No. NBP155066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FBXO11 Polyclonal specifically detects FBXO11 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation.

Specifications

Antigen FBXO11
Applications Western Blot, ChIP Assay
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation (ChIP)
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q86XK2
Gene Alias F-box only protein 11, F-box protein 11, FBX11MGC44383, FLJ12673, PRMT9VIT1protein arginine N-methyltransferase 9, ubiquitin protein ligase E3 component n-recognin 6, UBR6, VIT-1, Vitiligo-associated protein 1, vitiligo-associated protein VIT-1
Gene Symbols FBXO11
Host Species Rabbit
Immunogen Synthetic peptides corresponding to FBXO11(F-box protein 11) The peptide sequence was selected from the middle region of FBXO11 (NP_079409). Peptide sequence HDVEFIRHDRFFCDCGAGTLSNPCTLAGEPTHDTDTLYDSAPPIESNTLQ.
Molecular Weight of Antigen 94 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80204
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Amphibian, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.