Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ FBXO15, Mouse, Unlabeled, Clone: 3B11.1,

Catalog No. MABD127MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABD127MI 200 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD127MI Supplier MilliporeSigma™ Supplier No. MABD127
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

F-box only protein 15 (UniProt Q8NCQ5) is encoded by the FBXO15 (also known as FBX15) gene (Gene ID 201456) in human. Polyubiquitination of proteins is mediated by ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin ligases (E3s). There exist RING finger-type, HECT-type, U-box-type, and PHD-finger-type E3 ligases. The RING finger-type E3s include seven cullin-based E3s, such as SKP1–CUL1–F-box-protein (SCF) complex and the anaphase-promoting complex/cyclosome (APC/C). F-box proteins use their N-terminal F-box motif to bind Skp1 and other motif(s) in their C-terminal region for substrates recognition and recruitment. F-box proteins are divided into the FBXW subclass that contains WD40 repeat domains, the FBXL subclass that contains leucine-rich repeat domains, and the F-box only (FBXO) subclass that contains not yet fully characterized substrate recognition domains. SCF-Fbxo15 is reported to mediate the ubiquitination of cardiolipin synthase (CLS1) for proteasomal degradation in human lung epithelial cells. PTEN-induced putative kinase 1 (PINK1) is shown to phosphorylate CLS1 and cause its release from the inner mitochondrial membrane to the outer membrane, making it accessible for Fbxo15-targeted ubiquitination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO15
Applications Western Blot
Classification Monoclonal
Clone 3B11.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ antibody in buffer containing PBS with 0.05% Sodium Azide.
Gene FBXO15, FBX15
Gene Accession No. Q8NCQ5
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding an N-terminal sequence of human FBXO15.
Purification Method Purified by Ion-Exchange Chromatography
Quantity 200 μL
Regulatory Status RUO
Research Discipline Stem Cell Research
Primary or Secondary Primary
Gene ID (Entrez) NP_001136430
Test Specificity Clone 3B11.1 recoginzes an N-terminal epitope present in spliced isoform 1 (UniProt Q8NCQ5-1), but absent in spliced isoform 2 (UniProt Q8NCQ5-2).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.