Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528530 Supplier Invitrogen™ Supplier No. PA528530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji, Human ESC. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

F-box protein 15 (FBXO15) is a gene located on chromosome 20p11.1 that encodes a member of the F-box protein family, known for their role in ubiquitin-mediated proteolysis. Proteins in this family contain an F-box domain, enabling them to form part of the SCF (SKP1-CUL1-F-box protein) ubiquitin ligase complex, which targets specific proteins for ubiquitination and subsequent degradation. FBXO15 is involved in regulating various cellular processes by controlling the turnover of proteins related to cell cycle progression, signal transduction, and development. Its function is crucial for maintaining cellular homeostasis, where dysregulation of FBXO15 can impact protein degradation pathways, potentially leading to disorders such as cancer due to uncontrolled cell proliferation. Current research is focused on understanding the specific targets and functions of FBXO15 within the ubiquitin proteasome system and its potential as a therapeutic target in diseases characterized by aberrant proteolytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO15
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene FBXO15
Gene Accession No. Q8NCQ5
Gene Alias AU019763; ecat3; F-box only protein 15; F-box protein 15; FBX15; FBXO15; MGC39671
Gene Symbols FBXO15
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FBXO15. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201456
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.