Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fbxo16 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA562882
Description
Immunogen sequence: DPRSLCRCAQ VCWHWKNLAE LDQLWMLKCL RFNWYINFSP TPFEQGIWKK HYIQMVKELH ITKPKTPPKD GFVIADVQL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbx class.
Specifications
Fbxo16 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IX29 | |
FBXO16 | |
Recombit protein corresponding to Human Fbxo16 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
FBXO16 | |
FBX16, F-box only protein 16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
157574 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction