Learn More
Invitrogen™ FBXO2 Polyclonal Antibody

Description
FBXO2 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | FBXO2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | FBXO2 |
| Gene Accession No. | Q80UW2, Q9UK22 |
| Gene Alias | FBG1; F-box gene 1; F-box only protein 2; F-box protein 2; Fbs1; Fbs2; FBX2; FBXO2; neural F box protein NFB42; NFB42; OCP1; organ of Corti protein 1; prion protein ligand 4; Prpl4 |
| Gene Symbols | FBXO2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.