Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO21 Antibody (OTI2C7), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271711APC
Description
FBXO21 Monoclonal specifically detects FBXO21 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
| FBXO21 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| FBXO21 | |
| Full length recombinant protein of human FBXO21 (NP_296373) produced in HEK293T cells. | |
| 0.1 mL | |
| 23014.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2C7 | |
| Flow Cytometry | |
| DKFZp434G058, F-box only protein 21, F-box protein 21, FBX21FLJ90233, KIAA0875MGC26682 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction