Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO21 Monoclonal Antibody (OTI3A6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504049S


Description
FBXO21 Monoclonal antibody specifically detects FBXO21 in Human samples. It is validated for Western BlotSpecifications
FBXO21 | |
Monoclonal | |
0.65 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P23743 | |
Mouse | |
Affinity chromatography | |
RUO | |
1606 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI3A6 | |
Unconjugated | |
Dgka | |
2810425J22Rik; AU016673; F-box only protein 21; F-box protein 21; FBX21; FBXO21; KIAA0875; mKIAA0875 | |
Full length human recombinant protein of human DGKA produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction