Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO21 Monoclonal Antibody (OTI5H4), TrueMAB™, OriGene
SDP

Catalog No. 502540552
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0552 30 μL
50-167-4798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0552 Supplier OriGene Supplier No. TA504025S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FBXO21 Monoclonal antibody specifically detects FBXO21 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen FBXO21
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5H4
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Dgka
Gene Accession No. P23743, P51556
Gene Alias 2810425J22Rik; AU016673; F-box only protein 21; F-box protein 21; FBX21; FBXO21; KIAA0875; mKIAA0875
Host Species Mouse
Immunogen Full length human recombinant protein of human DGKA produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100855691, 140866, 1606
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.