Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO22 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $501.50
Specifications
Antigen | FBXO22 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15542520
![]() |
Novus Biologicals
NBP15542520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155425
![]() |
Novus Biologicals
NBP155425 |
100 μL |
Each for $501.50
|
|
|||||
Description
FBXO22 Polyclonal specifically detects FBXO22 in Human samples. It is validated for Western Blot.Specifications
FBXO22 | |
Polyclonal | |
Rabbit | |
Q8NEZ5 | |
26263 | |
Synthetic peptides corresponding to FBXO22(F-box protein 22) The peptide sequence was selected from the middle region of FBXO22. Peptide sequence CCKVGASNYLQQVVSTFSDMNIILAGGQVDNLSSLTSEKYVLCASDFVCE. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
F-box only protein 22, F-box protein 22, F-box protein FBX22p44, FBX22FLJ13986, FIST domain containing 1, FISTC1, MGC31799 | |
FBXO22 | |
IgG | |
30 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title