Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO22 Mouse anti-Human, Mouse, Clone: OTI1H1, TrueMAB™, Origene
SDP

Catalog No. 502243795
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-224-3795 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-224-3795 Supplier OriGene Supplier No. TA809766S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FBXO22 Monoclonal antibody specifically detects FBXO22 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen FBXO22
Applications Western Blot
Classification Monoclonal
Clone OTI1H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FBXO22
Gene Accession No. Q78JE5, Q8NEZ5
Gene Alias 0610033L19Rik; 1600016C16Rik; F-box only protein 22; F-box protein 22; F-box protein FBX22p44; Fbx22; Fbxo22; FIST domain containing 1; FISTC1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-220 of human FBXO22 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26263, 71999
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.