Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597025
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597025 Supplier Invitrogen™ Supplier No. PA597025
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YMADSETFIS LEECRGHKRA RKRTSMETAL ALEKLFPKQC QVLGIVTPGI VVTPMGSGSN RPQEIEIGES GFALLFPQIE GIKIQPFHFI KDPKNLTLER HQLTEVGLLD NPELRVVLVF GYNCCKVGAS NYLQQVVSTF SDMNIILAGG QVDNLSSLTS EKNPLDIDAS GVVGLSFSGH RIQSATVLLN EDVSDEKTAE AAMQRLKAAN IPEHNTIGFM FACVGRGFQY YRAKGNVEAD AFRKFFPSVP LFGFFGNGEI GCDRIVTGNF ILRKCNEVKD DDLFHSYTTI MALIHLGSSK; Positive Samples: HeLa, MCF-7; Cellular Location: Cytoplasm, Z line, myofibril, sarcomere.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Two transcript variants encoding different isoforms exist for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO22
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FBXO22
Gene Accession No. Q8NEZ5
Gene Alias 0610033L19Rik; 1600016C16Rik; F-box only protein 22; F-box protein 22; F-box protein FBX22p44; Fbx22; Fbxo22; FIST domain containing 1; FISTC1
Gene Symbols FBXO22
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 104-403 of human FBXO22 (NP_6717171).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26263
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.