Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO25 Monoclonal Antibody (OTI6F11)
GREENER_CHOICE

Catalog No. PIMA527287
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527287 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527287 Supplier Invitrogen™ Supplier No. MA527287
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FBXO25 Monoclonal Antibody for Western Blot, IHC (P)

FBXO25 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. FBXO25 belongs to the Fbxs class.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO25
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6F11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FBXO25
Gene Accession No. Q8TCJ0
Gene Alias 9130015I06Rik; AI649137; F-box only protein 25; F-box protein 25; F-box protein Fbx25; FBX25; FBXO25; MGC20256; MGC51975; Unknown (protein for MGC:137945); zgc:100907
Gene Symbols FBXO25
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-191 of FBXO25 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26260
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.