Learn More
FBXO25 Mouse anti-Human, Clone: OTI1A1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810350

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FBXO25 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. FBXO25 belongs to the Fbxs class.Specifications
FBXO25 | |
Monoclonal | |
Unconjugated | |
FBXO25 | |
FBX25 | |
Mouse | |
Affinity Chromatography | |
RUO | |
26260 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1A1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8TCJ0 | |
FBXO25 | |
Human recombit protein fragment corresponding to amino acids 1-191 of human FBXO25 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.