Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590255 Supplier Invitrogen™ Supplier No. PA590255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WQQQLQDLQM TKQVNNGLTL SDLPLHMLNN ILYRFSDGWD IITLGQVTPT LYMLSEDRQL WKKLCQYHFA EKQFCRHLIL SEKGHIEWKL MYFALQKHYP AKEQYGDTLH FCRHCSILFW KDSGHPCTAA DPDSCFTPVS PQHFIDLFKF; Positive Samples: DU145, 293T, Mouse testis; Cellular Location: Nucleus.

FBXO25 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. FBXO25 belongs to the Fbxs class.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO25
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FBXO25
Gene Accession No. Q8TCJ0, Q9D2Y6
Gene Alias 9130015I06Rik; AI649137; F-box only protein 25; F-box protein 25; F-box protein Fbx25; FBX25; FBXO25; MGC20256; MGC51975; Unknown (protein for MGC:137945); zgc:100907
Gene Symbols FBXO25
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 142-291 of human FBXO25 (NP_0363052).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26260, 66822
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.