Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO27 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017922 0.025 mg
NB10093569 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017922 Supplier Novus Biologicals Supplier No. NB10093569SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FBXO27 Polyclonal specifically detects FBXO27 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen FBXO27
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Alias Fbg5, FBG5FBX27, F-box only protein 27, F-box protein 27, F-box protein FBG5, F-box/G-domain protein 5, Fbx27
Gene Symbols FBXO27
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 163-175 (KKQVLDLEEEGLW) of the human FBG5 protein. 76% identical to mouse.
Molecular Weight of Antigen 30 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126433
Test Specificity FBG5
Target Species Rat, Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.