Learn More
Invitrogen™ FBXO27 Monoclonal Antibody (1G6)

Description
Peptide Sequence: WGARHDSGCM YRLLVQLLDA NQTVLDKFSA VPDPIPQWNN NACLHVTHVF SNIKMGVRFV SFEHRGQDTQ FWAGHYGARV TNSSVIVRVR LS.
Specifications
Specifications
| Antigen | FBXO27 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1G6 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Fbxo27 |
| Gene Accession No. | Q8NI29 |
| Gene Alias | E130008B10Rik; F Box Gama 5; FBG5; F-box only protein 27; F-box protein 27; F-box protein FBG5; F-box/G-domain protein 5; Fbx27; FBXO27; Gm161; rCG54033-like; RGD1563982 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.