Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXO27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566475
Description
Immunogen sequence: ARSCQSPARNA RPCPLGRFCA RRPIGRNLIR NPCGQEGLRK WMVQHGGDG Highest antigen sequence identity to the following orthologs - mouse 76%, rat 66%.
F-box proteins, a critical component of the evolutionary conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a third subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.
Specifications
FBXO27 | |
Polyclonal | |
Unconjugated | |
Fbxo27 | |
E130008B10Rik; F Box Gama 5; FBG5; F-box only protein 27; F-box protein 27; F-box protein FBG5; F-box/G-domain protein 5; Fbx27; FBXO27; Gm161; rCG54033-like; RGD1563982 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126433 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NI29 | |
Fbxo27 | |
Recombinant Human FBXO27. Recombinant protein control fragment (Product #RP-107148). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction