Learn More
Invitrogen™ FBXO28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114048
Description
Immunogen sequence: KVQEQQKQLQ DQDQKLLEQT QIIGEQNARL AELERKLREV MESAVGNSSG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 75%; Zebrafish: 91%.
Members of the F-box protein family, such as FBXO28, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.
Specifications
| FBXO28 | |
| Polyclonal | |
| Unconjugated | |
| FBXO28 | |
| 4833428J17Rik; 5730505P19Rik; CENP-30; centromere protein 30; D1Ertd578e; F-box only protein 28; F-box protein 28; Fbx28; FBXO28; KIAA0483; mKIAA0483 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23219 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NVF7 | |
| FBXO28 | |
| Synthetic peptide directed towards the middle region of human FBXO28. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.