Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109713 Supplier Invitrogen™ Supplier No. PA5109713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CKRMDLVCQR MLNQGFLKVE RYHNLCQKQV KAQLPRRESE RRNHSLARHA DILAAVETRL SLLNMTFMKY VDSNLCCFIP GKVIDEIYRV LRYVNSTRAP QRAHEVLQEL RDISSMAMEY FDEKIVPILK RKLPGSDVSG RLMGSPPVPG PSAALTTMQL FSKQNPSRQE VTKLQQQVKT NGAGVTVLRR EISELRTKVQ EQQKQLQDQD QKLLEQTQII GE.

Members of the F-box protein family, such as FBXO28, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO28
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FBXO28
Gene Accession No. Q9NVF7
Gene Alias 4833428J17Rik; 5730505P19Rik; CENP-30; centromere protein 30; D1Ertd578e; F-box only protein 28; F-box protein 28; Fbx28; FBXO28; KIAA0483; mKIAA0483
Gene Symbols FBXO28
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 91-312 of human FBXO28 (NP_055991.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23219, 305105
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.