Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542299 Supplier Invitrogen™ Supplier No. PA542299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NCCYVSRRLS QLSSHDPLWR RHCKKYWLIS EEEKTQKNQC WKSLFIDTYS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

FBXO3 encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FBXO3
Gene Accession No. Q9UK99
Gene Alias 1200002G09Rik; 1700026K02Rik; AI046358; DKFZp564B092; FBA; F-box only protein 3; F-box protein 3; F-box protein FBA; F-box protein FBX3; FBX3; FBXO3
Gene Symbols FBXO3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FBXO3 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26273
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.