Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXO3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564886
Description
Immunogen sequence: IGCKLPDDYRC SYRIHNGQKL VVPGLLGSMA LSNHYRSEDL LDVDTAAGGF QQRQGLKYCL PLTFCIHTGL SQYIAVEAAE GRNKNEVFYQ CPDQMARNPA AIDMFIIGAT FTDWFTSYVK NVVSGGFPII RDQIFRYVHD PECV Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
FBXO3 encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs.
Specifications
FBXO3 | |
Polyclonal | |
Unconjugated | |
FBXO3 | |
1200002G09Rik; 1700026K02Rik; AI046358; DKFZp564B092; FBA; F-box only protein 3; F-box protein 3; F-box protein FBA; F-box protein FBX3; FBX3; FBXO3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UK99 | |
FBXO3 | |
Recombinant Human FBXO3. Recombinant protein control fragment (Product #RP-104544). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction