Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113522
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113522 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113522 Supplier Invitrogen™ Supplier No. PA5113522
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FBXO3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

FBXO3 encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FBXO3
Gene Accession No. Q9DC63, Q9UK99
Gene Alias 1200002G09Rik; 1700026K02Rik; AI046358; DKFZp564B092; FBA; F-box only protein 3; F-box protein 3; F-box protein FBA; F-box protein FBX3; FBX3; FBXO3
Gene Symbols FBXO3
Host Species Rabbit
Immunogen Recombinant Human F-box only protein 3 protein (276-376AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26273, 57443
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.