Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO30 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318377
This item is not returnable.
View return policy
Description
FBXO30 Polyclonal antibody specifically detects FBXO30 in Human samples. It is validated for ImmunoprecipitationSpecifications
FBXO30 | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
84085 | |
Store at 4°C. Do not freeze. | |
IgG |
Immunoprecipitation | |
1 mg/ml | |
Immunoprecipitation 8 - 15 ug/mg lysate | |
F-box domain protein, F-box only protein 30, F-box only protein, helicase, 18, F-box protein 30, FBX30, FLJ41030, MGC21674 | |
Recognizes region between residues Between Between 200 and 250.. | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction