Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564944 Supplier Invitrogen™ Supplier No. PA564944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APSFNFLSNSC WSKPKEDKAV DTSDLEVAED PMGLQGIDLI TAALLFCLGD SPGGRGISDS RMADIYHIDV GTQTFSLPSA ILATSTMVGE IASASACDHA NPQLSNPSPF QTLGLDLVLE CVARYQPKQR SMFTFVCGQL FRRKEF Highest antigen sequence identity to the following orthologs - mouse 94%, rat 93%.

Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. Required for muscle atrophy following denervation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO30
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXO30
Gene Accession No. Q8TB52
Gene Alias 1700026A16Rik; F-box domain protein; F-box only protein 30; F-box only protein, helicase, 18; F-box protein 30; Fbx30; Fbxo30; MUSA1; Muscle ubiquitin ligase of SCF complex in atrophy-1
Gene Symbols FBXO30
Host Species Rabbit
Immunogen Recombinant Human FBXO30. Recombinant protein control fragment (Product #RP-105724).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.