Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO30 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260712
Description
FBXO30 Polyclonal specifically detects FBXO30 in Human samples. It is validated for Immunoprecipitation.Specifications
FBXO30 | |
Polyclonal | |
Immunoprecipitation 8-15 ul/mg lysate | |
F-box domain protein, F-box only protein 30, F-box only protein, helicase, 18, F-box protein 30, FBX30, FLJ41030, MGC21674 | |
Rabbit | |
Affinity Purified | |
Primary | |
Human | |
IgG |
Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
FBXO30 | |
between 200 and 250 | |
100 μL | |
84085 | |
Store at 4degreesC. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction