Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO31 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119088F
Description
FBXO31 Polyclonal antibody specifically detects FBXO31 in Human, Mouse samples. It is validated for ImmunofluorescenceSpecifications
FBXO31 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
DKFZp434B027, F-box only protein 31, F-box protein 31, FBX14DKFZp434J1815, Fbx31, FBXO14FLJ22477, MGC15419, MGC9527, pp2386, putative breast cancer tumor-suppressor, SCF ubiquitin ligase specificity factor | |
The immunogen recognized by this antibody maps to a region between residue 489 and 539 of human F-box protein 31 using the numbering given in entry NP_079011.3 (GeneID 79791). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Immunofluorescence | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79791 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction