Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO31 Antibody (OTI1H1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272208PE
Description
FBXO31 Monoclonal specifically detects FBXO31 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
FBXO31 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
FBXO31 | |
Full length recombinant protein of human FBXO31 (NP_079011) produced in HEK293T cells. | |
0.1 mL | |
79791.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI1H1 | |
Flow Cytometry | |
DKFZp434B027, F-box only protein 31, F-box protein 31, FBX14DKFZp434J1815, Fbx31, FBXO14FLJ22477, MGC15419, MGC9527, pp2386, putative breast cancer tumor-suppressor, SCF ubiquitin ligase specificity factor | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction