Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO31 Monoclonal Antibody (1C5)
GREENER_CHOICE

Catalog No. PIMA524553
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524553 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524553 Supplier Invitrogen™ Supplier No. MA524553
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: GQGQPFVLPV GVSSRNEDYP RTCRMCFYGT GLIAGHGFTS PERTPGVFIL FDEDRFGFVW LELKSFSLYS RVQATFRNAD APSPQAFDEM LKNIQSLTS.

Members of the F-box protein family, such as FBXO31, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO31
Applications ELISA
Classification Monoclonal
Clone 1C5
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FBXO31
Gene Accession No. Q5XUX0
Gene Alias 1110003O08Rik; 2310046N15Rik; DKFZp434B027; DKFZp434J1815; F-box only protein 14; F-box only protein 31; F-box protein 31; FBX14; FBX31; FBXO14; Fbxo31; FLJ22477; MGC15419; MGC9527; MNCb-2609; MRT45; pp2386; putative breast cancer tumor-suppressor; RGD1561069; SCF ubiquitin ligase specificity factor
Gene Symbols FBXO31
Host Species Mouse
Immunogen FBXO31 (NP_079011.3, 269 a.a. ~ 367 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79791
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.