Learn More
Invitrogen™ FBXO31 Monoclonal Antibody (OTI1A11)

Description
FBXO31 Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | FBXO31 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI1A11 |
| Concentration | 1.32 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FBXO31 |
| Gene Accession No. | Q5XUX0 |
| Gene Alias | 1110003O08Rik; 2310046N15Rik; DKFZp434B027; DKFZp434J1815; F-box only protein 14; F-box only protein 31; F-box protein 31; FBX14; FBX31; FBXO14; Fbxo31; FLJ22477; MGC15419; MGC9527; MNCb-2609; MRT45; pp2386; putative breast cancer tumor-suppressor; RGD1561069; SCF ubiquitin ligase specificity factor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.