Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588839 Supplier Thermo Scientific Supplier No. PA588839
Add to Cart
Add to Cart

FBXO31 Polyclonal Antibody

FBXO31 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO31
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FBXO31
Gene Accession No. B2RYN2, Q3TQF0
Gene Alias 1110003O08Rik; 2310046N15Rik; DKFZp434B027; DKFZp434J1815; F-box only protein 14; F-box only protein 31; F-box protein 31; FBX14; FBX31; FBXO14; Fbxo31; FLJ22477; MGC15419; MGC9527; MNCb-2609; MRT45; pp2386; putative breast cancer tumor-suppressor; RGD1561069; SCF ubiquitin ligase specificity factor
Gene Symbols FBXO31
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 240-539 of human FBXO31 (NP_0790113).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498959, 76454
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.